4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
4-Dimethylaminocinnamaldehyde, 98%
Thermo Scientific Chemicals
4-Dimethylaminocinnamaldehyde, 98%
CAS: 6203-18-5 | C11H13NO | 175.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA115790250
Price (MYR)
1,458.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,458.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Dimethylaminocinnamaldehyde
Name Note98%
CAS6203-18-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning