3-Dimethylaminobenzoic acid, 99%
3-Dimethylaminobenzoic acid, 99%
3-Dimethylaminobenzoic acid, 99%
3-Dimethylaminobenzoic acid, 99%
Thermo Scientific Chemicals
3-Dimethylaminobenzoic acid, 99%
CAS: 99-64-9 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA115750250
Price (MYR)
540.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
540.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Dimethylaminobenzoic acid
Name Note99%
CAS99-64-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning