4-Dimethylaminoantipyrine, 97%
4-Dimethylaminoantipyrine, 97%
4-Dimethylaminoantipyrine, 97%
4-Dimethylaminoantipyrine, 97%
4-Dimethylaminoantipyrine, 97%
CAS: 58-15-1 | C13H17N3O | 231.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA115740250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Dimethylaminoantipyrine
CAS58-15-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0