Dimethyl acetylenedicarboxylate, 98%
Dimethyl acetylenedicarboxylate, 98%
Dimethyl acetylenedicarboxylate, 98%
Dimethyl acetylenedicarboxylate, 98%
Thermo Scientific Chemicals
Dimethyl acetylenedicarboxylate, 98%
CAS: 762-42-5 | C6H6O4 | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA115711000
Price (MYR)
1,248.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,248.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethyl acetylenedicarboxylate
CAS762-42-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
View more
Alkylation reagent
RUO – Research Use Only