2,2-Dimethoxypropane, 98+%
2,2-Dimethoxypropane, 98+%
2,2-Dimethoxypropane, 98+%
2,2-Dimethoxypropane, 98+%
Thermo Scientific Chemicals
2,2-Dimethoxypropane, 98+%
CAS: 77-76-9 | C5H12O2 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA115635000
Price (MYR)
401.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
401.00
BOT
Add to cart
Specifications
Chemical Name or Material2,2-Dimethoxypropane
CAS77-76-9
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Alkylation reagent
RUO – Research Use Only