Dimethoxymethane, 99.5+%
Dimethoxymethane, 99.5+%
Dimethoxymethane, 99.5+%
Dimethoxymethane, 99.5+%
Thermo Scientific Chemicals
Dimethoxymethane, 99.5+%
CAS: 109-87-5 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA115560010
Price (MYR)
466.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
466.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethoxymethane
CAS109-87-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Store in a well-ventilated place.
Keep container tightly closed.