3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
Thermo Scientific Chemicals
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%
CAS: 2478-38-8 | C10H12O4 | 196.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA115540250
Price (MYR)
2,763.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,763.00
EA
Add to cart
Specifications
Chemical Name or Material3′,5′-Dimethoxy-4′-hydroxyacetophenone
Name Note97%
CAS2478-38-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning