3,4-Dimethoxybenzyl alcohol, 96%
3,4-Dimethoxybenzyl alcohol, 96%
3,4-Dimethoxybenzyl alcohol, 96%
3,4-Dimethoxybenzyl alcohol, 96%
3,4-Dimethoxybenzyl alcohol, 96%
CAS: 93-03-8 | C9H12O3 | 168.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA115491000
Price (MYR)
1,056.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,056.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4-Dimethoxybenzyl alcohol
CAS93-03-8
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0