2,4-Dimethoxybenzaldehyde, 98%
2,4-Dimethoxybenzaldehyde, 98%
2,4-Dimethoxybenzaldehyde, 98%
2,4-Dimethoxybenzaldehyde, 98%
Thermo Scientific Chemicals
2,4-Dimethoxybenzaldehyde, 98%
CAS: 613-45-6 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA115380250
Price (MYR)
309.00
G
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
309.00
G
Add to cart
Specifications
Chemical Name or Material2, 4-Dimethoxybenzaldehyde
CAS613-45-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0