3,5-Dimethoxyaniline, 98%
3,5-Dimethoxyaniline, 98%
3,5-Dimethoxyaniline, 98%
3,5-Dimethoxyaniline, 98%
Thermo Scientific Chemicals
3,5-Dimethoxyaniline, 98%
CAS: 10272-07-8 | C8H11NO2 | 153.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA115360250
Price (MYR)
731.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
731.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 5-Dimethoxyaniline
Name Note98%
CAS10272-07-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning