2,3-Dimercapto-1-propanol, 97.0-100.5%
2,3-Dimercapto-1-propanol, 97.0-100.5%
2,3-Dimercapto-1-propanol, 97.0-100.5%
2,3-Dimercapto-1-propanol, 97.0-100.5%
Thermo Scientific Chemicals
2,3-Dimercapto-1-propanol, 97.0-100.5%
CAS: 59-52-9 | C3H8OS2 | 124.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA115300250
Price (MYR)
3,846.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,846.00
EA
Add to cart
Specifications
Chemical Name or Material2,3-Dimercapto-1-propanol
Name Note97.0-100.5%
CAS59-52-9
CAS Min %97.0
Health Hazard 1GHS Signal Word: Danger