N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
N,N-Diisopropylethylamine, 99+%
Thermo Scientific Chemicals
N,N-Diisopropylethylamine, 99+%
CAS: 7087-68-5 | C8H19N | 129.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA115225000
Price (MYR)
1,444.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,444.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN,N-Diisopropylethylamine
Name Note98+%
CAS7087-68-5
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger