1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
Thermo Scientific Chemicals
1,6-Diisocyanatohexane, 99+%
CAS: 822-06-0 | C8H12N2O2 | 168.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA115191000
Price (MYR)
434.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
434.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 6-Diisocyanatohexane
Name Note99+%
CAS822-06-0
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger