1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
1,6-Diisocyanatohexane, 99+%
Thermo Scientific Chemicals
1,6-Diisocyanatohexane, 99+%
CAS: 822-06-0 | C8H12N2O2 | 168.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA115190250
Price (MYR)
422.00
Bottle
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
422.00
Bottle
Specifications
Chemical Name or Material1, 6-Diisocyanatohexane
Name Note99+%
CAS822-06-0
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger