4,4'-Diiodobiphenyl, 99%
4,4'-Diiodobiphenyl, 99%
4,4'-Diiodobiphenyl, 99%
4,4'-Diiodobiphenyl, 99%
Thermo Scientific Chemicals
4,4'-Diiodobiphenyl, 99%
CAS: 3001-15-8 | C12H8I2 | 406 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA115120250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4, 4'-Diiodobiphenyl
CAS3001-15-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0