2,7-Dihydroxynaphthalene, 97%
2,7-Dihydroxynaphthalene, 97%
2,7-Dihydroxynaphthalene, 97%
2,7-Dihydroxynaphthalene, 97%
Thermo Scientific Chemicals
2,7-Dihydroxynaphthalene, 97%
CAS: 582-17-2 | C10H8O2 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA115045000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 7-Dihydroxynaphthalene
CAS582-17-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0