3,4-Dihydroxycinnamic acid, 99+%, predominantly trans isomer
CAS: 331-39-5 | C9H8O4 | 180.16 g/mol
Have Questions?
Change view
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA114930250
Price (MYR)
1,074.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,074.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4-Dihydroxycinnamic acid
CAS331-39-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0