3,4-Dihydroxybenzoic acid, 97%
3,4-Dihydroxybenzoic acid, 97%
3,4-Dihydroxybenzoic acid, 97%
3,4-Dihydroxybenzoic acid, 97%
Thermo Scientific Chemicals
3,4-Dihydroxybenzoic acid, 97%
CAS: 99-50-3 | C7H6O4 | 154.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA114891000100 gGlass bottle
Catalog number FSA114891000
Price (MYR)
1,479.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,479.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4-Dihydroxybenzoic acid
Name Note97%
CAS99-50-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning