2,4-Dihydroxybenzoic acid, 97%
2,4-Dihydroxybenzoic acid, 97%
2,4-Dihydroxybenzoic acid, 97%
2,4-Dihydroxybenzoic acid, 97%
Thermo Scientific Chemicals
2,4-Dihydroxybenzoic acid, 97%
CAS: 89-86-1 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA114875000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Dihydroxybenzoic acid
Name Note97%
CAS89-86-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning