2',5'-Dihydroxyacetophenone, 97%
2',5'-Dihydroxyacetophenone, 97%
2',5'-Dihydroxyacetophenone, 97%
2',5'-Dihydroxyacetophenone, 97%
Thermo Scientific Chemicals
2',5'-Dihydroxyacetophenone, 97%
CAS: 490-78-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA114810250
Price (MYR)
609.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
609.00
BOT
Add to cart
Specifications
Chemical Name or Material2', 5'-Dihydroxyacetophenone
Name Note97%
CAS490-78-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning