3,4-Dihydro-2H-pyran, 99%
3,4-Dihydro-2H-pyran, 99%
3,4-Dihydro-2H-pyran, 99%
3,4-Dihydro-2H-pyran, 99%
Thermo Scientific Chemicals
3,4-Dihydro-2H-pyran, 99%
CAS: 110-87-2 | C5H8O | 84.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA114781000100 gGlass bottle
Catalog number FSA114781000
Price (MYR)
307.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
307.00
EA
Add to cart
Specifications
Chemical Name or Material3, 4-Dihydro-2H-pyran
CAS110-87-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0