Diglycolic anhydride, 97%
Diglycolic anhydride, 97%
Diglycolic anhydride, 97%
Diglycolic anhydride, 97%
Thermo Scientific Chemicals
Diglycolic anhydride, 97%
CAS: 4480-83-5 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA114671000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiglycolic anhydride
Name Note0.97
CAS4480-83-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement