1,4-Difluorobenzene, 99+%
1,4-Difluorobenzene, 99+%
1,4-Difluorobenzene, 99+%
1,4-Difluorobenzene, 99+%
Thermo Scientific Chemicals
1,4-Difluorobenzene, 99+%
CAS: 540-36-3 | C6H4F2 | 114.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA114610250
Price (MYR)
597.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
597.00
EA
Add to cart
Specifications
Chemical Name or Material1, 4-Difluorobenzene
Name Note99+%
CAS540-36-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger