2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
Thermo Scientific Chemicals
2,4-Difluoroaniline, 99%
CAS: 367-25-9 | C6H5F2N | 129.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number FSA114590100
Price (MYR)
322.00
BOT
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
322.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4-Difluoroaniline
CAS367-25-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0