N,N-Diethyl-m-toluamide, 98%
N,N-Diethyl-m-toluamide, 98%
N,N-Diethyl-m-toluamide, 98%
N,N-Diethyl-m-toluamide, 98%
Thermo Scientific Chemicals
N,N-Diethyl-m-toluamide, 98%
CAS: 134-62-3 | C12H17NO | 191.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA114571000
Price (MYR)
246.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
246.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Diethyl-m-toluamide
Name Note98%
CAS134-62-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning