N,N-Diethylhydroxylamine, 97%
N,N-Diethylhydroxylamine, 97%
N,N-Diethylhydroxylamine, 97%
N,N-Diethylhydroxylamine, 97%
Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%
CAS: 3710-84-7 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA114425000
Price (MYR)
714.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
714.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Diethylhydroxylamine
Name Note97%
CAS3710-84-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning