7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
7-Diethylamino-4-methylcoumarin, 99%
Thermo Scientific Chemicals
7-Diethylamino-4-methylcoumarin, 99%
CAS: 91-44-1 | C14H17NO2 | 231.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA114050050
Price (MYR)
278.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
278.00
EA
Add to cart
Specifications
Chemical Name or Material7-Diethylamino-4-methylcoumarin
Name Note99%
CAS91-44-1
CAS Min %98.5
CAS Max %100.0