View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Diethylaminoethylchloride hydrochloride
CAS869-24-9
CAS Min %99.4
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0