4-(Diethylamino)benzaldehyde, 99%
4-(Diethylamino)benzaldehyde, 99%
4-(Diethylamino)benzaldehyde, 99%
4-(Diethylamino)benzaldehyde, 99%
Thermo Scientific Chemicals
4-(Diethylamino)benzaldehyde, 99%
CAS: 120-21-8 | C11H15NO | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA114001000
Price (MYR)
384.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
384.00
BOT
Add to cart
Specifications
Chemical Name or Material4-(Diethylamino)benzaldehyde
Name Note99%
CAS120-21-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger