1,4-Dicyanobenzene, 98%
1,4-Dicyanobenzene, 98%
1,4-Dicyanobenzene, 98%
1,4-Dicyanobenzene, 98%
Thermo Scientific Chemicals
1,4-Dicyanobenzene, 98%
CAS: 623-26-7 | C8H4N2 | 128.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA113821000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Dicyanobenzene
CAS623-26-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0