1,2-Dichloropropane, 98%, pure
1,2-Dichloropropane, 98%, pure
1,2-Dichloropropane, 98%, pure
1,2-Dichloropropane, 98%, pure
Thermo Scientific Chemicals
1,2-Dichloropropane, 98%, pure
CAS: 78-87-5 | C3H6Cl2 | 112.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA113670010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Dichloropropane
Name Note98%
CAS78-87-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger