2,6-Dichlorophenol, 99%
2,6-Dichlorophenol, 99%
2,6-Dichlorophenol, 99%
2,6-Dichlorophenol, 99%
Thermo Scientific Chemicals
2,6-Dichlorophenol, 99%
CAS: 87-65-0 | C6H4Cl2O | 163.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA113590250
Price (MYR)
305.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
305.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 6-Dichlorophenol
Name Note99%
CAS87-65-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger