Check Order Status
3,4-Dichloronitrobenzene, 95%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
3,4-Dichloronitrobenzene, 95%, Thermo Scientific™
Have Questions?
Change view
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number
FSA113532500
Price (MYR)
301.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
301.00
BOT
Add to cart
Specifications
Chemical Name or Material
3, 4-Dichloronitrobenzene
Name Note
95%
CAS
99-54-7
CAS Min %
94.0
Health Hazard 1
GHS Signal Word:
Danger