2,3-Dichloro-1,4-naphthoquinone, 98%
2,3-Dichloro-1,4-naphthoquinone, 98%
2,3-Dichloro-1,4-naphthoquinone, 98%
2,3-Dichloro-1,4-naphthoquinone, 98%
Thermo Scientific Chemicals
2,3-Dichloro-1,4-naphthoquinone, 98%
CAS: 117-80-6 | C10H4Cl2O2 | 227.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA113480250
Price (MYR)
293.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
293.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 3-Dichloro-1, 4-naphthoquinone
Name Note98%
CAS117-80-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning