Dichlorodiphenylsilane, 97%
Dichlorodiphenylsilane, 97%
Dichlorodiphenylsilane, 97%
Dichlorodiphenylsilane, 97%
Thermo Scientific Chemicals
Dichlorodiphenylsilane, 97%
CAS: 80-10-4 | C12H10Cl2Si | 253.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA113341000
Price (MYR)
450.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
450.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichlorodiphenylsilane
CAS80-10-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit