2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%
Thermo Scientific Chemicals
2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%
CAS: 84-58-2 | C8Cl2N2O2 | 227.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
Glass bottle
Catalog number FSA113301000
Price (MYR)
2,073.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,073.00
BOT
Add to cart
Specifications
Chemical Name or Material2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
CAS84-58-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Oxidation reagent
RUO – Research Use Only