1,2-Dichlorobenzene, 99%, pure
1,2-Dichlorobenzene, 99%, pure
1,2-Dichlorobenzene, 99%, pure
1,2-Dichlorobenzene, 99%, pure
Thermo Scientific Chemicals
1,2-Dichlorobenzene, 99%, pure
CAS: 95-50-1 | C6H4Cl2 | 147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number FSA113180250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Dichlorobenzene
CAS95-50-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0