2,6-Dichlorobenzaldehyde, 99%
2,6-Dichlorobenzaldehyde, 99%
2,6-Dichlorobenzaldehyde, 99%
2,6-Dichlorobenzaldehyde, 99%
Thermo Scientific Chemicals
2,6-Dichlorobenzaldehyde, 99%
CAS: 83-38-5 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA113162500
Price (MYR)
1,465.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,465.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 6-Dichlorobenzaldehyde
Name Note99%
CAS83-38-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger