3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
3,4-Dichloroaniline, 98%
Thermo Scientific Chemicals
3,4-Dichloroaniline, 98%
CAS: 95-76-1 | C6H5Cl2N | 162.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA113110050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 4-Dichloroaniline
Name Note98%
CAS95-76-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger