2,6-Dichloroaniline, 98%
2,6-Dichloroaniline, 98%
2,6-Dichloroaniline, 98%
2,6-Dichloroaniline, 98%
Thermo Scientific Chemicals
2,6-Dichloroaniline, 98%
CAS: 608-31-1 | C6H5Cl2N | 162.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA113100250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Dichloroaniline
Name Note98%
CAS608-31-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger