Dichloroacetic acid, 99+%
Dichloroacetic acid, 99+%
Dichloroacetic acid, 99+%
Dichloroacetic acid, 99+%
Thermo Scientific Chemicals
Dichloroacetic acid, 99+%
CAS: 79-43-6 | C2H2Cl2O2 | 128.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA113062500
Price (MYR)
923.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
923.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichloroacetic acid
CAS79-43-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Suspected of causing cancer.
May damage fertility or the unborn child.
May cause harm to breast-fed children.
May cause damage to or
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat