2,6-Di-tert-butylphenol, 99%
2,6-Di-tert-butylphenol, 99%
2,6-Di-tert-butylphenol, 99%
2,6-Di-tert-butylphenol, 99%
Thermo Scientific Chemicals
2,6-Di-tert-butylphenol, 99%
CAS: 128-39-2 | C14H22O | 206.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA113000010
Price (MYR)
452.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
452.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 6-Di-tert-butylphenol
CAS128-39-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0