Di-n-butylamine, 99%
Di-n-butylamine, 99%
Di-n-butylamine, 99%
Di-n-butylamine, 99%
Thermo Scientific Chemicals
Di-n-butylamine, 99%
Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL | CAS: 111-92-2 | C8H19N | 129.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
1 L
2.5 L
Packaging:
View all
Glass bottle
Catalog number FSA112950500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-butylamine
CAS111-92-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre