alpha,alpha'-Dibromo-p-xylene, 97%
alpha,alpha'-Dibromo-p-xylene, 97%
alpha,alpha'-Dibromo-p-xylene, 97%
alpha,alpha'-Dibromo-p-xylene, 97%
Thermo Scientific Chemicals
alpha,alpha'-Dibromo-p-xylene, 97%
CAS: 623-24-5 | C8H8Br2 | 263.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA112940250
Price (MYR)
533.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
533.00
BOT
Add to cart
Specifications
Chemical Name or Materialα,α′-Dibromo-p-xylene
Name Note97%
CAS623-24-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger