2,5-Dibromonitrobenzene, 97%
2,5-Dibromonitrobenzene, 97%
2,5-Dibromonitrobenzene, 97%
2,5-Dibromonitrobenzene, 97%
Thermo Scientific Chemicals
2,5-Dibromonitrobenzene, 97%
CAS: 3460-18-2 | C6H3Br2NO2 | 280.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA112861000
Price (MYR)
868.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
868.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 5-Dibromonitrobenzene
Name Note97%
CAS3460-18-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning