1,6-Dibromohexane, 98%
1,6-Dibromohexane, 98%
1,6-Dibromohexane, 98%
Thermo Scientific Chemicals
1,6-Dibromohexane, 98%
CAS: 629-03-8 | C6H12Br2 | 243.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA112815000
Price (MYR)
1,071.00
Bottle
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,071.00
Bottle
Specifications
Chemical Name or Material1,6-Dibromohexane
Name Note98%
CAS629-03-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning