2,4'-Dibromoacetophenone, 98%
2,4'-Dibromoacetophenone, 98%
2,4'-Dibromoacetophenone, 98%
2,4'-Dibromoacetophenone, 98%
Thermo Scientific Chemicals
2,4'-Dibromoacetophenone, 98%
CAS: 99-73-0 | C8H6Br2O | 277.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA112680100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4'-Dibromoacetophenone
CAS99-73-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0