Dibenzothiophene, 98%
Dibenzothiophene, 98%
Dibenzothiophene, 98%
Dibenzothiophene, 98%
Thermo Scientific Chemicals
Dibenzothiophene, 98%
CAS: 132-65-0 | C12H8S | 184.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA112541000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDibenzothiophene
CAS132-65-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
Collect spillage.
Dispose of contents/