1,4-Diazabicyclo[2.2.2]octane, 97%
1,4-Diazabicyclo[2.2.2]octane, 97%
1,4-Diazabicyclo[2.2.2]octane, 97%
1,4-Diazabicyclo[2.2.2]octane, 97%
Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%
CAS: 280-57-9 | C6H12N2 | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number FSA112475000
Price (MYR)
1,287.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,287.00
EA
Add to cart
Specifications
Chemical Name or Material1,4-Diazabicyclo[2.2.2]octane
Name Note97%
CAS280-57-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger