3,4-Diaminotoluene, 97%
3,4-Diaminotoluene, 97%
3,4-Diaminotoluene, 97%
3,4-Diaminotoluene, 97%
Thermo Scientific Chemicals
3,4-Diaminotoluene, 97%
CAS: 496-72-0 | C7H10N2 | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA112431000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3, 4-Diaminotoluene
CAS496-72-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0